Learn More
Thermo Scientific™ Orion™ ISE Calibration Standards
Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.
Brand:  Thermo Scientific™ 941707
 View more versions of this product
Product Code. 10292391
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.
All standards are 1 pint (475mL) bottles, unless otherwise noted.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chloride Standard
475 mL
C of A Acceptance Range: 100.0 ppm +/- 0.5 ppm Sodium Chloride as Chloride
Chloride Ion Selective Electrodes Including 9617BNWP, 9417BN, 9417SC
Colorless
Calibration Standard
1 Bottle
Yes
Yes
100 ppm as Cl-
Calcium Ion Selective Electrode
Calibration Standard
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberDZ1 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog Number941707
Lot NumberCW1 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number941707
Lot NumberCZ1 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number941707
Lot NumberBZ1 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog Number941707
Lot NumberAT1 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number941707
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ ISE Calibration Standards > 100ppm as Cl-; 475mL; CAS: (7647-14-5)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.