Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Product Code. 11782208
Click to view available options
Type:
Aluminum
Ammonia
Chloride
Chlorine
Chlorine Dioxide
Copper
Iron
Manganese
Nitrate
Nitrite
Ozone
Phosphate
Silica
Sulfate
Sulfide
Zinc
Range:
0.04 to 0.25 mg/L Al
0.08 to 1 mg/L N
0.2 to 1.5 mg/L N
0.2 to 3 mg/L N
0.2 to 3 mg/L S
0.2 to 5 mg/L O3
0.2 to 6 mg/L Fe
0.3 to 3 mg/L Zn
0.3 to 8 mg/L PO4
0.4 to 5 mg/L Cl2
0.5 to 10 mg/L SiO2
0.5 to 12 mg/L Cu
0.5 to 7 mg/L N
0.8 to 11 mg/L ClO2
1.0 to 7.5 mg/L N
1.5 to 14 mg/L N
10 to 100 mg/L SO4
2 to 30 mg/L Mn
2.5 to 40 mg/L Cl-
5 to 50 mg/L NO3
Method:
Bariumsulfate-Turbidity
Bathocuproine
Cadmium Reduction
DPD
Diazotization (Azo)
Direct Nesslerization
Eriochrome Cyanine R
Ferric Thiocyanide
Heteropoly Blue
Methylene Blue
Periodate
Phenanthroline
Salicylate
Stannous Chloride
Zincon
Unit Size:
30 tests
Pack of 30
Product Code. 11782208
Brand: Thermo Scientific™ AC4078
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors
TRUSTED_SUSTAINABILITY
Specifications
Type Iron
Test Reagent Formats Ampoule
For Use With (Equipment) Orion AQUAfast AQ4000 Colorimeter
Description AQUAfast iron (total & soluble) Auto-ID ampoules, 30 pack
Includes 30 Auto-ID Ampoules
Method Phenanthroline
For Use With (Application) Colorimetric water and wastewater testing equipment
Recommended Accessories AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER)
Range 0.2 to 6 mg/L Fe
EPA Approved Methods Wastewater: Standard Methods 3500-Fe-1997
Quantity 30 Ampoules
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.