Learn More
B Braun™ Original Perfusor™ Syringes with Aspiration Needle
Use this 3-piece syringe for infusion pumps. B Braun™ Original Perfusor™ Syringes with Aspiration enable slow patient during infusion of drug solutions.
€ 2.00 - € 146.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10346534
|
B Braun™
8728810F |
50 mL |
€ 2.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10294072
|
B Braun™
8728810F |
50 mL |
€ 146.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10106073
|
B Braun™
8728623 |
20 mL |
€ 123.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Polypropylene
- High contrast graduation
- Permanent markings for readability
- Graduation in ml increments