Learn More
Macherey-Nagel™ Optima™ WAX GC Capillary Column
Product Code. 12482840 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
2 μm
Unit Size:
Each
Product Code. 12482840
Brand: Macherey-Nagel™ 726549.25
Only null left
Specifications
Diameter (Metric) 0.8 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.53 mm
Stationary Phase Optima™ WAX
USP Type G16
For Use With (Application) Aqueous Solutions Analysis, Aqueous Solutions
Temperature Range 240°C to 250°C
Film Thickness 1 μm
Length (Metric) 25 m
Quantity 1 Each