Learn More
Macherey-Nagel™ Optima™ FFAP GC Capillary Column
Brand: Macherey-Nagel™ 726116.25
This item is not returnable.
View return policy
Specifications
0.4 mm | |
0.25 mm | |
G1, G2, G38 | |
240°C to 250°C | |
25 m |
GC Capillary Column | |
Optima™ FFAP | |
Free Carboxylic Acids Analysis, Free Carboxylic Acids | |
0.25 μm | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ FFAP GC Capillary Column > Length: 25m; I.D.: 0.25mm; Film Thickness: 0.25um