Learn More
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column
Brand: Macherey-Nagel™ 726320.30
This item is not returnable.
View return policy
Specifications
0.5 mm | |
0.32 mm | |
G46 | |
300°C to 320°C | |
30 m |
High Performance GC Capillary Column | |
Optima™ 1701 | |
Gas Chromatography (GC) | |
0.5 μm | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ 1701 High Performance GC Capillary Column > Length: 30m; I.D.: 0.32mm; Film Thickness: 0.5um