Learn More
Macherey-Nagel™ Optima™ 1301 GC Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
1 μm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 0.8 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | Optima™ 1301 |
| USP Type | G1, G2, G38 |
| For Use With (Application) | Pesticide Analysis |
| Temperature Range | 300°C to 320°C |
| Film Thickness | 1 μm |
| Length (Metric) | 25 m |
| Quantity | 1 Each |