Learn More
Macherey-Nagel™ Optima™ 1 Capillary Column for GC
Brand: Macherey-Nagel™ 726038.15
Specifications
0.4 mm | |
0.25 mm | |
G1, G2, G38 | |
340°C to 360°C | |
15 m |
GC Capillary Column | |
Optima™ 1 | |
Gas Chromatography (GC) | |
0.1 μm | |
1 Each |