Learn More
Lovibond™ W600 Optical Glass Cells
Click to view available options
Path Length:
25 mm
50 mm
Unit Size:
Each
Description
- Optical glass cells
- Suitable for use in a variety of Lovibond™ colorimeters
Specifications
Specifications
| Type | Cell |
| Material | Optical Glass |
| Quantity | 1 Each |
| Path Length | 25 mm |
| For Use With (Equipment) | Colorimetry |