Learn More
Köttermann Open Mobile Cabinet
Click to view available options
No. of Drawers:
2
3
None
No. of Shelves:
1
None
Dimensions (D x W x H):
500 x 450 x 590 mm
500 x 450 x 740 mm
500 x 600 x 590 mm
500 x 600 x 740 mm
500 x 900 x 590 mm
500 x 900 x 740 mm
516 x 450 x 590 mm
516 x 600 x 590 mm
500 x 1200 x 590 mm
500 x 1200 x 740 mm
missing translation for 'unitSize'
Each
Description
- Versatile storage space
- Mobile with lockable wheels
- Open-front design provides easy access to stored items
- Many sizes and configurations available
Specifications
Specifications
| Color | White |
| Cabinet Type | Mobile |
| Dimensions (D x W x H) | 500 x 1200 x 740 mm |
| Door Type | None |
| Finish | Epoxy Coated |
| Frame Material | Steel |
| No. of Doors | None |
| No. of Drawers | None |
| No. of Shelves | None |
| Warranty | None |
| Show More |