Learn More
Diba Omnifit™ Replacement Endpieces for EZ Columns
Click to view available options
Endcapped:
Adjustable
Fixed
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Unit Size:
Each
Specifications
Specifications
| Endcapped | Fixed |
| For Use With (Equipment) | Omnifit™ EZ Columns |
| Diameter (Metric) Inner | 10 mm |
| Type | Liquid Chromatography Column |
| Length (Metric) | 4 cm |
| Material | PTFE |