Learn More
Ohaus™ OIML F1 Weight Set
Adjusted to Meet or Exceed OIML F1 Tolerances
Brand:  Ohaus™ 80780414
 View more versions of this product
Product Code. 16622472
  • € 872.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Weights Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free to protect your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Certificate
1.269 kg
Protective Case
Stainless Steel, Aluminum
2 x 200 g to 1 mg
23/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers