missing translation for 'onlineSavingsMsg'
Learn More

3M™ Oil Sorbent Sheets, T-151

High capacity petroleum sorbent pads have a scrim on one side to increase strength and help eliminate linting. Conveniently sized pads allow users to quickly spread sorbent over, and in the path of a spill.

Brand:  3M™ 7000073485

Product Code. 17148640

  • € 364.00 / Pack of 200
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for absorbing oil from areas where water may also be present
  • Handy dispenser box for easy on-site use
  • Each box contains 100 pads (430 x 480 mm)
  • Sorbs up to 140 liters in total
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brown
140 L
T-151
450 x 450 mm
450 mm
39.63 gal.
Oil Sorbent Sheet
100/Cs.
450 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers