Learn More
Welch Ilmvac™ Oil
Brand: Welch Ilmvac™ 800122
Specifications
Oil | |
Rotary Vane Pumps | |
1 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Welch Ilmvac™ Oil > Volume: 1L