missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Obinutuzumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541839
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Obinutuzumab is a humanized anti-CD20 monoclonal antibody, originated by GlycArt Biotechnology AG and developed by Roche as a cancer treatment. It was approved under the trade name Gazyva by the US FDA in 2013, and as Gazyvaro by the EMA in Europe, for the treatment of chronic lymphocytic leukemia in combination with chemotherapy in treatment-naive patients, and as a second line treatment for follicular lymphoma. Obinutuzumab is a fully humanized monoclonal antibody that binds to an epitope on CD20 that partially overlaps with the epitope recognized by rituximab. Obinutuzumab binds to CD20 on B cells and causes these cells to be destroyed by engaging the adaptive immune system, directly activating intracellular apoptosis pathways, and activating the complement system.Specifications
Obinutuzumab | |
Recombinant Monoclonal | |
Unconjugated | |
afutuzumab; Gazyva; Gazyvaro; RG7159; RO5072759 | |
Human MS4A1/CD20 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
3.54 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction