Learn More
Masterflex™ Nylon T Connector
Brand:  Masterflex™ 30613-22
 View more versions of this product
Product Code. 12334628
  • € 31.75 / Pack of 10
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Specifications
12.7 mm
12.7mm
10 Pack
121°C
Nylon
-46°C
T
T-Connector
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Masterflex™ Nylon T Connector > Fits Tubing: 12.7mm