Learn More
Masterflex™ Nylon T Connector
Product Code. 12304628
Click to view available options
For Use With:
4.76mm
6.35mm
9.52mm
12.7mm
19.05mm
25.4mm
Unit Size:
Pack of 10
Product Code. 12304628
Brand: Masterflex™ 3061310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Diameter (Metric) Inner 4.76 mm
Material Nylon
For Use With 4.76mm
Min. Operating Temperature (Metric) -46°C
Packaging Quantity 10 Pack
Shape T
Max. Operating Temperature (Metric) 121°C
Product Type T-Connector