Learn More
Masterflex™ Nylon Sanitary Clamp Fittings
Product Code. 16629882
Click to view available options
Size (English) Grip:
2 in.
4 in.
Unit Size:
Each
Product Code. 16629882
Brand: Masterflex™ 3050829
Only null left
Add to basket
Add to basket
Compatible with Metal and Plastic Tri-Clamp™ Fittings
  • Excellent chemical resistance
  • Low leachable characteristics
  • Autoclavable
Specifications
Autoclavable Autoclavable
Sterilization Method Autoclave or Ethylene Oxide (ETO)
Type Sanitary Clamp
Material Nylon
For Use With (Equipment) Tri-Clamp Fittings
Size (English) Grip 4 in.
Color White