Learn More
Masterflex™ Nylon Sanitary Clamp Fittings
Click to view available options
Size (English) Grip:
2 in.
4 in.
Unit Size:
Each
Description
- Excellent chemical resistance
- Low leachable characteristics
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Sterilization Method | Autoclave or Ethylene Oxide (ETO) |
| Type | Sanitary Clamp |
| Material | Nylon |
| For Use With (Equipment) | Tri-Clamp Fittings |
| Size (English) Grip | 2 in. |
| Color | White |