missing translation for 'onlineSavingsMsg'
Learn More

NUP98-NUP96 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen NUP98-NUP96
Concentration 0.27 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16879983
View Documents
Proteintech
12329-1-AP-20UL
20 μL
€ 145.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16869983
View Documents
Proteintech
12329-1-AP-150UL
150 μL
€ 410.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody was raised against the carboxy-terminal region of Nup98-Nup96, it recognizes all Nup98-Nup96 proteins except Nup98-N. Since the autoproteolysis is very rapid this antibody usually detected 98-115 kDa Nup96 protein in multiple cell lines. It

ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
Specifications

Specifications

NUP98-NUP96
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Rat, Mouse, Human
P49793, P52948, Q6PFD9
269966, 4928, 81738
NUP98-NUP96 Fusion Protein Ag3016
Primary
-20°C
NUP98
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
96 kDa nucleoporin, ADAR2, ADIR2, Nucleoporin Nup96, NUP196, NUP96, NUP98 NUP96, NUP98-NUP96
NUP98
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.