Learn More
Thermo Scientific™ Nunc™ Rectangular Dishes
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Brand: Thermo Scientific™ 267060
Description
Non-treated Thermo Scientific™ Nunc™ Rectangular Dishes with ANSI™ standard dimensions are optimized for molecular biology applications using robotic instruments.
Rectangular Dishes 1, 4 and 8-Well
- 1-well dish is ideal for HTS Petri dish applications
- 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
- 4-well dish holds four glass slides for storage
1-Well Format
- Convenient plate format for cell culture
- HTS Petri dish applications
4- and 8-Well Formats
- Assay compartmentalization
- Probe multiple gene subsets
- Wash Western blot strips
- 4-well dish holds four glass slides
- Useful in all areas of cell culture, including scale-up and cloning
Ordering Information:
Customized bar coding is available
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
1 | |
Non-treated | |
Rectangular | |
100 | |
100 mm |
Cell Culture Dish | |
Sterile | |
90 mL | |
Lid | |
10 | |
86 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1419889 | Certificate of Processing | 31/01/2025 | 267060 |
1419773 | Certificate of Processing | 31/01/2025 | 267060 |
1415670 | Certificate of Processing | 02/01/2025 | 267060 |
1419889 | Certificate of Compliance | 18/01/2025 | 267060 |
1419773 | Certificate of Compliance | 17/01/2025 | 267060 |