Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
Description
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
- Molded of durable PETG
- Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
- Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
- Reduces wrist strain and increases productivity with quick-action ergonomic closure
- Can be traced by lot number on every bottle
- Cell culture expansion
- Vaccine production
- Antibody production
Specifications
Specifications
Bottle Surface | Pleated |
Color | Clear |
Description | 4260-22 |
Material | PETG |
No. per Case | 22 |
No. per Pack | 22 |
Packaging | 22/case |
Cell Growth Area | 4200 cm2 |
Closure Included | Yes |
Graduated | Yes |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Documents
Scientific Resources
Application Notes
Brochures
Product Information
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1418298 | Certificate of Processing | 02/01/2025 | 4260-22 |
1418155 | Certificate of Processing | 30/12/2024 | 4260-22 |
1418298 | Certificate of Compliance | 13/12/2024 | 4260-22 |
1305549 | Certificate of Compliance | 09/12/2024 | 4260-22 |
1285149 | Certificate of Compliance | 09/12/2024 | 4260-22 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.