Learn More
Thermo Scientific™ Nunc™ General Long Term Storage Tubes
Product Code. 10726662
Click to view available options
Volume (Metric):
1.8 mL
4.5 mL
Unit Size:
Pack of 1200
Pack of 1800
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 10726662
Marque: Thermo Scientific™ 347783
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Contain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Contiain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific™ Nunc™ General Long Term Storage Tubes.
Starfoot vial fits into grooves of the CryoTube Rack (Cat. No. 376589), which allows one hand operation.
  • Caps and tubes packaged separately to save handling time
  • Tubes are nonsterile
  • Economical choice for less demanding applications
TRUSTED_SUSTAINABILITY
Spécification
Description Nunc Stor-It Tubes, external thread, round bottom
Color White
Diameter (Metric) Outer 48 mm
Sterility Non-sterile
Volume (Metric) 1.8 mL
No. per Case 1800
No. per Pack 450
Style Starfoot, round-bottom
Volume (Metric) Total 1.8 mL
Height (Metric) 12.5 mm
Quantity 1800/Cs., 450/Pk.
Type Storage Tube