Learn More
Thermo Scientific™ Nunc™ Containers
Click to view available options
Capacity (Metric):
12.5 mL
20 mL
30 mL
50 mL
Unit Size:
Pack of 1000
Pack of 1200
Pack of 2400
Pack of 30
Pack of 800
Description
Store samples safely using Thermo Scientific™ Nunc™ Containers. Nunc Labware Products are made from high purity resins and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Features:
- Transparent polystyrene construction
- Versatile containers made of high-quality resins
- Snap caps available for all containers (not included)
- Nonsterile (except for 561301)
Specifications
Specifications
| Product Type | Container |
| Closure Included | No |
| Material | Polystyrene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Capacity (Metric) | 30 mL |
| Closure Type | Snap Cap |
| Color | Clear |