missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates
Click to view available options
Color:
Black
Natural
White
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Unit Size:
Pack of 100
Pack of 25
Description
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.
Features:
- Rounded wells are optimized for mixing and sample recovery
- Square rounded wells minimize wicking (capillary action)
- Natural plates for storage identification or storing colorimetric assays
- White plates for luminescent assays
- Black plates for fluorescent assays
- Customized bar coding is available
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | Certified RNase and DNase Free. ANSI Compliant. |
| Format | Standard |
| Lid | Without Lid |
| Description | 384-well Polypropylene Sample Processing & Storage Plates |
| Array | 16 x 24 |
| Autoclavable | Autoclavable |
| Packaging | 25 pk., 100 cs. |
| Binding Type | Low binding PP |
| Product Line | Nunc |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction