missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 RNA Blood, 96-well kit for RNA from blood

Product Code. p-7159596
Click to view available options
No. of Reactions:
2 x 96
4 x 96
Unit Size:
192 reactions
384 reactions
This item is not returnable. View return policy

Product Code. 15809067

Brand: Macherey-Nagel™ 740225.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete kit with proven 96-well format for high throughput isolation of total RNA from fresh or frozen whole blood (human or animal) via direct total blood lysis that requires no selective erythrocyte lysis.

NucleoSpin™ 96 RNA Blood Midi kit achieves superior RNA yield and quality in a format that provides convenient handling and flexible sample volume and throughput.

  • Direct total blood lysis – very simple and convenient procedure
  • No selective erythrocyte lysis at 4 °C – complete processing at RT
  • Compatible with common blood collection tubes and anticoagulants (e.g., EDTA, citrate, and heparin)
  • Suitable for fresh and frozen blood samples
  • rDNase included for efficient removal of genomic DNA
  • Processing under vacuum or by centrifugation
  • Suitable for manual and automated (vacuum or centrifugation) processing
  • Innovative MN Wash Plate minimizes risk of cross-contamination

Specifications

Format 96-well Plate
Isolation Technology Silica Membrane Technology
Elution Volume 5 to 130 μL
Sample Type Whole Blood (Fresh or Frozen)
Efficiency Binding capacity: 100 μg
Final Product Type Total RNA
For Use With (Equipment) Centrifuge or Vacuum
For Use With (Application) RNA isolation
Includes NucleoSpin RNA Blood Binding Plates, MN Wash Plates, Square-well Blocks, Round-well Blocks Low, Elution Plates U-bottom, Self-adhering Foil, buffers, RNase-free rDNase, Liquid Proteinase K
No. of Reactions 2 x 96
Purity A260/A280: 1.9 - 2.1
Test Time 100 min./Plate
Volume (Metric) < 400 μL
Yield 1 to 8 μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.