Learn More
Nucleoside-Analog-2, MedChemExpress
MedChemExpress Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.
Brand: Medchem Express HY-77652
Description
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.Specifications
Nucleoside-Analog-2 | |
Gray | |
C9H11N5O6 | |
DMSO : 33.33 mg/mL (116.86 mM; Need ultrasonic) ∣H2O : 20 mg/mL (70.12 mM; Need ultrasonic) | |
285.21 | |
95.0% | |
Solid |
876708-01-9 | |
Research | |
50 mg | |
O[C@@H]1[C@](CO)(N=[N+]=[N-])O[C@@H](N2C(NC(C=C2)=O)=O)[C@H]1O | |
285.21 | |
Research | |
COVID-19-anti-virus |
Product Suggestions
Customers who viewed this item also viewed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only