Learn More
Macherey-Nagel™ Nucleogel™ SUGAR 810 H HPLC Columns
Product Code. 10378251 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Each
Product Code. 10378251
Brand: Macherey-Nagel™ 719574
Only null left
Specifications
Length (Metric) 30 mm
Type HPLC Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOGEL™ SUGAR 810 H
Column Format Valco HPLC Column
For Use With (Application) Separation of Sugar and Organic Acids
Technique HPLC
USP Type L17