Learn More
Macherey-Nagel™ Nucleodur™ C18 Isis Analytical HPLC EC Column
Product Code. p-8012212 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
50 mm
75 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2 mm
3 mm
4 mm
4.6 mm
Unit Size:
Each
Product Code. 12468447
Brand: Macherey-Nagel™ 760412.20
Only null left
Specifications
Column Chemistry Silica
Column Format Analytical Column
Crosslinking Polymeric octadecyl
Diameter (Metric) 2 mm
For Use With HPLC
Length (Metric) 125 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility Acetonitrile/water
Particle Size 5 μm
pH 1 to 10
Pore Size 110 Å
Quantity 1 Ea.
Solvents Acetonitrile/water
Surface Area 340 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18 Isis
Packing Material Silica
Carbon Load 20%
Column Type Steric Selectivity
Endcapped Yes
Phase Reversed Phase
Type Analytical EC Column