Learn More
Invitrogen™ Streptavidin Agarose
Product Code. 11558976
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. 11558976
Brand: Invitrogen™ SA10004
Includes
Streptavidin Agarose is supplied as 10mL of a 50% slurry containing 5mL of packed gel bed in 0.01M sodium phosphate (pH 7.2), 0.15M NaCl, and 0.02% sodium azide.
For isolating biotinylated molecules and complexes with biotinylated components, including antibodies, antibody-antigen complexes, ligands and receptors, nucleic acids, and nucleic acid-protein complexes
  • Especially useful as an immunoprecipitation reagent for antibodies that do not bind well to protein A
  • The matrix consists of streptavidin covalently attached to cross-linked 4% agarose beads through a 15-atom spacer arm
  • Streptavidin Agarose can bind 3 to 8mg of biotinylated IgG/mL of gel
Immunoprecipitation, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10 mg/mL
Quantity 5 mL
Content And Storage Streptavidin Agarose is supplied as 10 ml of a 50% slurry containing 5 ml of packed gel bed in 0.01 M sodium phosphate (pH 7.2), 0.15 M NaCl, and 0.02% sodium azide. Store at 4°C.
Form Liquid Suspension
Product Type Streptavidin Agarose