missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Non-treated 384-well Microplates
Manufactured without surface treatment to create low biomolecular binding. Greiner Bio One™ Non-treated 384-Well Assay Microplates are ideal for sensitive biochemical screening applications.
€ 501.00 - € 856.00
Specifications
Certifications/Compliance | DIN ISO 9001 |
---|---|
Format | Standard |
Array | 16 x 24 |
Binding Type | Medium Binding |
Material | Polystyrene |
Product Code | Brand | Color | Quantity | Lid | Total Volume (Metric) Well | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Lid | Total Volume (Metric) Well | Price | Quantity & Availability | ||
10527071
|
Greiner Bio-One
781096 |
Black | 40/Cs. | Without Lid | 131 μL |
€ 856.00
Pack of 40 |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||
10060202
|
Greiner Bio-One
781101 |
Clear | 100/Cs. | Without Lid | 131 μL |
€ 501.00
Pack of 100 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
||
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Quadrupled the number of wells (from 96)
- Well-to-well spacing: 4.5 mm
- Rounded square well profile, providing flexible working volume of 10 μl to 130 μl and reduced wicking and bubbling
Specifications
DIN ISO 9001 | |
16 x 24 | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) |
Standard | |
Medium Binding | |
384 | |
Non-treated |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title