Learn More
Fisherbrand™ Non-Slip Rubber Mat
Non-Slip Rubber Mat for Multi-Platform Shaker
Brand: Fisherbrand™ 88861130
38.54 EUR valid until 2025-07-31
Use promo code "24166" to get your promotional price.
Description
- For use with 88861021 and 88861022 Multi-Platform Shakers
Specifications
306 x 246 mm | |
88861021, 88861022 |
Promotions
-
Fisherbrand
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24166Expires: 2025-07-31
-
Multiple Manufacturers
Illuminating Protein Pathways
Shining a light on the tools that drive protein discovery.Promo Code: 24148Expires: 2025-08-22
New Offer Save Now