Learn More
NM107, MedChemExpress
MedChemExpress NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.
Brand: Medchem Express HY-10468
This item is not returnable.
View return policy
Description
NM107 (2'-C-Methylcytidine) is an nucleoside inhibitor of the hepatitis C virus (HCV) NS5B polymerase, the EC50 of NM107 in the wild-type replicon cells is 1.85 μM.Specifications
NM107 | |
White | |
C10H15N3O5 | |
2'-C-Methylcytidine NM-107 | |
NC(C=CN1[C@@H]2O[C@H](CO)[C@@H](O)[C@@]2(C)O)=NC1=O | |
257.24 | |
Research | |
COVID-19-anti-virus |
20724-73-6 | |
Research | |
50 mg | |
DMSO : 100 mg/mL (388.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (194.37 mM) | |
257.24 | |
98.9% | |
Solid |
Research purposes only