Learn More
Schmidlin™ NM Plus Rack Hydrogen Generator
This generator produces a continuous supply of high purity, dry hydrogen.
Brand:  Schmidlin™ DB-RNM1000-EU
Encompass_Preferred
 View more versions of this product
Product Code. 17708929
  • € 24465.29 / Each
Explore more special offers
Description
Description
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Pressure: 11 barg (160 psig)
  • Unique permeation membrane drying system
  • Cold dual dynamic regeneration dryer
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Meets and exceeds requirements for the most demanding GC applications
  • Superior hydrogen production with reliable long life cell
  • Minimal maintenance; no desiccant cartridges to change
  • PC monitoring for maintenance, diagnostics and remote connection
Specifications
Specifications
500 cm
13.3 cm
11 bar
48.5 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
400 W
1000 mL/min.
35°C
19 in.
Outlet: 1/8 in.
>99.99996%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers