missing translation for 'onlineSavingsMsg'
Learn More

Schmidlin™ NM Plus FID Station H2 and Zero Air Generator

This dual gas generator produces a continuous supply of high purity hydrogen and zero air.

Brand:  Schmidlin™ DB-FNM300-EU

Encompass_Preferred

 View more versions of this product

Product Code. 17788879

  • € 18558.81 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Increase efficiency and improve performance
  • On-demand supply 24/7
  • Zero air <0.1 ppm
  • THC Pressure: 11 barg (160 psig)
  • Permeation membrane drying system
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC and GC/MS applications
Specifications

Specifications

68 cm
15 cm
11 bar
69 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
150 W
300 mL/min.
35°C
27 in.
Outlet: 1/8 in.
>99.99996%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers