Learn More
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with NS Stopcock
Click to view available options
Standard Taper Joint:
NS 14/23 Cone
NS 14/23 Socket
Unit Size:
Each
Description
- With conical ground joint and NS stopcock
- Made of DURAN™ tubing
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Duran Glass |
| Standard Taper Joint | NS 14/23 Cone |