missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene-d5, for NMR, 99 atom % D

Product Code. p-172593 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
Unit Size:
10g
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10752011

Brand: Thermo Scientific Acros 166360050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 4165-60-0 | C6H5NO2 | 128.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-60-0
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 128.14
MDL Number MFCD00044415
InChI Key LQNUZADURLCDLV-RALIUCGRSA-N
Synonym nitrobenzene-d5, benzene-d5, nitro, nitrobenzene d5, benzene-d5-, nitro, 1-nitro 2 h? benzene, nitrobenzene-d5, 99.5 atom % d, nitro 2h5 benzene, 1-nitro 2h5 benzene, 2h5 benzene, nitro, nitrobenzene-d5, analytical standard
PubChem CID 123210
IUPAC Name 1,2,3,4,5-pentadeuterio-6-nitrobenzene
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])[N+]([O-])=O

Specifications

Physical Form Liquid
Color Colorless to Yellow
Deuterium (D) 98.5% min.
Packaging Glass Bottle
Quantity 5 g
Linear Formula C6D5NO2
Formula Weight 128.15
Grade NMR
Chemical Name or Material Nitrobenzene-d5, 99 atom % D
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nitrobenzene-d5
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H351-Suspected of causing cancer.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.