Learn More
Hach Lange™ Nitrite Cuvette Tests
Product Code. 10267792
Click to view available options
Range:
0.015 to 0.6 mg/L NO2-N, 0.05 to 2.0 mg/L NO2
0.6 to 6mg/L NO3-N
Unit Size:
0.3kg
Product Code. 10267792
Brand: Hach Lange™ LCK341
Only null left
Add to basket
Add to basket
Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.
  • Nitrite cuvette test 0.015- 0.6 mg/L NO2-N
  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • EN ISO 26777, DIN 38405 D10 certifications
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents
  • Diazotisation method
  • Shelf life of 23 months from production date
  • Measuring range: 0.015 – 0.6 mg/L NO2-N
  • Measuring range (2): 0.05 - 2 mg/L NO2
Specifications
No. of Tests 25
Range 0.015 to 0.6 mg/L NO2-N, 0.05 to 2.0 mg/L NO2
Storage Requirements 15°C to 25°C
Test Method Diazotization
Type Nitrite Cuvette Test
compliance-icons
Product Identifier
  • Nitrat/Nitrate, Sample cuvette
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Citric acid