Learn More
Nitazoxanide-d4, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.
Specifications
Specifications
| Chemical Name or Material | Nitazoxanide-d4 |
| CAS | 1246819-17-9 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C12H5D4N3O5S |
| Color | Off-White |
| Synonym | NTZ-d4 NSC-697855-d4 |
| SMILES | O=C(NC1=NC=C([N+]([O-])=O)S1)C2=C(OC(C)=O)C([2H])=C([2H])C([2H])=C2[2H] |
| Molecular Weight (g/mol) | 311.31 |
| Quantity | 10 mg |
| Show More |
Research purposes only