Learn More
Nipecotic acid, 98%
Product Code. 11476406
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11476406
Brand: Thermo Scientific Alfa Aesar B24723.06
Request Bulk or Custom Format
CAS: 498-95-3 | C6H11NO2 | 129.16 g/mol
GABA reuptake inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 498-95-3
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00005992 MFCD01630787
InChI Key XJLSEXAGTJCILF-UHFFFAOYSA-N
Synonym nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine
PubChem CID 4498
ChEBI CHEBI:116931
IUPAC Name piperidine-3-carboxylic acid
SMILES OC(=O)C1CCCNC1
Specifications
Melting Point ∼261°C (decomposition)
Quantity 5 g
Beilstein 81096
Merck Index 14,6561
Solubility Information Soluble in water
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material Nipecotic acid