Learn More
Cole-Parmer™ Nickel/Stainless Steel Lab Spatulas
Product Code. 12363800 Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
16.51 cm
18.1 cm
20.32 cm
Unit Size:
Pack of 3
Product Code. 12363800
Brand: Cole-Parmer™ 0636903
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for precise sampling of powders or salts. Nickel/stainless steel spatulas come in a variety of end styles.
  • Nickel/SS Lab Spatulas
  • Available with a glazed or mirror finish
  • Easy to clean
  • Multiple sizes and tyles to fit a variety of laboratory needs
  • Pack of 3
Specifications
Quantity 3 Pk.
Product Type Laboratory Spatula
Length (English) 6.5 in.
Material Stainless Steel (304), Nickel
Length (Metric) 16.51 cm