Learn More
Cole-Parmer™ Nickel-plated Brass Luer Fittings
Brand: Cole-Parmer™ 41507-62
This item is not returnable.
View return policy
Specifications
Autoclavable | |
Male Luer Lock | |
1 Pack | |
Luer Fitting |
Nickle-plated Brass | |
Harsh chemical environments | |
1/16 in. Hose Barb |
Product Content Correction
Product Title
Cole-Parmer™ Nickel-plated Brass Luer Fittings > Connections: Male Luer Lock; Includes 1/16 in. Hose Barb