Learn More
Usbeck Laborgeraete™ Nickel Plated Brass Hoffman Pinchcocks
Click to view available options
Width (Metric):
17 mm
20 mm
25 mm
30 mm
40 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | Each |
| Type | Hoffman Pinchcock |
| Material | Nickel-plated Brass |
| Width (Metric) | 25 mm |