Learn More
Emma Safety Footwear New York Safety Shoes
Product Code. 15978431
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15978431
Brand: Emma Safety Footwear MM43264744
Only null left
Add to basket
Add to basket
Light and comfortable with knitted textile upper and Nano Carbon Composite protective nose. Ideal for workplaces where safety and long-lasting comfort are top priorities. Integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimum breathability.
  • S1P-Safety shoe with sole plate for dry work
  • Knitted Textile
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Gray, Black
Design New York
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 44
Toe Material Composite