Learn More
Smeg™ Liquid Acid Neutralizers
Achieve excellent washing results and optimize the thermal disinfection of laboratory instruments with these detergents. Smeg™ Liquid Acid Neutralizers are especially designed to guarantee efficient cleaning and optimize the efficiency of the final thermal disinfection phase.
Brand: Smeg™ ACIDGLP2
Description
- Acid liquid neutralizing agent
- For laboratory glassware
- 5 liter can
ACIDGLASS P2: organic phosphoric acid neutralizer
- pH 2.5
- Application field: general chemistry, inorganic and petrol chemistry
- 1.1 gallon / 5 liter container, 4.4x10-3-6.6x10-3gallon/liter / 2-3 ml/liter
ACIDGLASS C2: weak organic acid neutralizer
- pH 2.4
- Application field :universal, environmental analysis
- 1.1 gallon / 5 liter container, 2.2x10-3,4.4x10-3 gallon/liter / 1-2 ml/liter
Specifications
Acidglass P2 | |
Organic phosphoric acid neutralizer | |
1 Ea. |
Canister | |
General chemistry, inorganic and petrol chemistry | |
5 L |
Safety and Handling
- ACIDGLASS P2
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.