missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCLN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534356
This item is not returnable.
View return policy
Description
A suggested positive control is mouse heart tissue lysate. PA5-34356 can be used with blocking peptide PEP-1399.
May antagonize Nodal signaling and subsequent organization of axial structures during mesodermal patterning. [UniProt].
Specifications
| NCLN | |
| Polyclonal | |
| Unconjugated | |
| Ncln | |
| 3100002P13Rik; AW120537; NCLN; NET59; Nicalin; nicalin homolog; nicalin homolog (zebrafish); nicastrin-like protein; RGD1309355 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103425, 314648, 56926 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q5XIA1, Q8VCM8, Q969V3 | |
| Ncln | |
| An 18 amino acid synthetic peptide near the carboxy terminus of human NCLN. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction