Learn More
Abeba™ Nature 4045 Shoes
Leather covered cork footbed is ESD dissipative.
Brand: Abeba™ 4045/45
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- Night blue
- Slip resistance special rubber outsole sole
- Leather coated cork footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Adjustable instep area
- Buckle fastening
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Cork | |
45 | |
Rubber |
Night Blue | |
Leather | |
Slip-resistant | |
4045 |