Learn More
Abeba™ Nature 4045 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 4045/48
 View more versions of this product
Product Code. 15621348
  • € 98.90 / 1 pair
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • Night blue
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
48
Rubber
Night Blue
Leather
Slip-resistant
4045