Learn More
mb2 Stool on Glides - Wood - Mid Lift
Click to view available options
:
Holds 5 Pads
Description
- Stool with wood seat
- Pneumatic cylinder for height adjustment
- 5-star reinforced nylon base 640 mm dia.
- Glides
Specifications
Specifications
| Product Type | Stool |
| Color | Natural |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Autoclavable | Not Autoclavable |
| Finish | Fireproof |
| Height (Metric) | 646 mm |
| Holds | 5 Pads |
| Seating Height | 450 to 630 mm |
| Weight (Metric) | 5.78 kg |
| Show More |