Learn More
Thermo Scientific Chemicals Septum 16mm (15.9mm real) natural rubber
Shop All Thermo Scientific Chemicals Products
Click to view available options
Diameter (Metric):
1.59 cm
3.07 cm
Quantity:
100/Ea.
50/Ea.
Unit Size:
Pack of 100
Pack of 50
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Diameter (Metric) | 3.07 cm |
| Quantity | 100/Ea. |
| Color | Natural |
| Material | Rubber |
| Product Type | Rubber Septum |